Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165375.3 | NP_724335.2 | 1 - 2 | 295 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |