Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169587.2 | NP_001163058.1 | 6 - 7 | 2008 | 60 | |
NM_001169588.2 | NP_001163059.1 | 6 - 7 | 1068 | 60 | ||
NM_078897.4 | NP_523621.2 | 3 - 4 | 564 | 60 | ||
NM_165445.4 | NP_724428.1 | 6 - 7 | 2011 | 60 | ||
GenBank mRNA | AY069815.1 | - | 6 - 7 | 1996 | 60 | |
BT005201.1 | - | 6 - 7 | 1247 | 60 | ||
M92841.1 | - | 6 - 7 | 1580 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |