Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273790.1 | NP_001260719.1 | 3 - 4 | 392 | 58 | |
NM_001273791.1 | NP_001260720.1 | 3 - 4 | 392 | 58 | ||
NM_136359.3 | NP_610203.2 | 3 - 4 | 404 | 58 | ||
NM_165458.2 | NP_724453.1 | 3 - 4 | 483 | 58 | ||
NM_165459.2 | NP_724454.1 | 2 - 3 | 534 | 58 | ||
NM_165460.2 | NP_724455.2 | 3 - 4 | 404 | 58 | ||
GenBank mRNA | BT011368.1 | - | 3 - 4 | 413 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |