Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298200.1 | NP_001285129.1 | 5 - 6 | 1340 | 55 | |
NM_078570.3 | NP_511125.2 | 5 - 6 | 1340 | 55 | ||
GenBank mRNA | AY050238.1 | - | 5 - 6 | 1294 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |