Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103497.2 | NP_001096967.1 | 5 - 6 | 1891 | 64 | |
NM_078589.4 | NP_511144.2 | 4 - 5 | 1573 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |