Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103504.2 | NP_001096974.1 | 3 - 4 | 1166 | 73 | |
NM_078592.3 | NP_511147.2 | 3 - 4 | 1385 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |