Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103504.2 | NP_001096974.1 | 5 - 6 | 2219 | 61 | |
NM_001169274.2 | NP_001162745.1 | 4 - 5 | 1592 | 61 | ||
NM_001169275.2 | NP_001162746.1 | 4 - 5 | 1811 | 61 | ||
NM_001169276.3 | NP_001162747.1 | 4 - 5 | 1592 | 61 | ||
NM_001272597.2 | NP_001259526.2 | 4 - 5 | 1811 | 61 | ||
NM_001298290.1 | NP_001285219.1 | 5 - 6 | 1617 | 61 | ||
NM_078592.3 | NP_511147.2 | 5 - 6 | 2438 | 61 | ||
GenBank mRNA | AF195496.1 | - | 4 - 5 | 1445 | 61 | |
BT015267.1 | - | 4 - 5 | 1445 | 61 | ||
BT132967.1 | - | 4 - 5 | 1584 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |