Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103504.2 | NP_001096974.1 | 7 - 8 | 2612 | 61 | |
NM_001169274.2 | NP_001162745.1 | 6 - 7 | 1985 | 61 | ||
NM_001169275.2 | NP_001162746.1 | 6 - 7 | 2204 | 61 | ||
NM_001169276.3 | NP_001162747.1 | 6 - 7 | 1985 | 61 | ||
NM_001272597.2 | NP_001259526.2 | 6 - 7 | 2204 | 61 | ||
NM_001298290.1 | NP_001285219.1 | 7 - 8 | 2010 | 61 | ||
NM_078592.3 | NP_511147.2 | 7 - 8 | 2831 | 61 | ||
GenBank mRNA | AF195496.1 | - | 6 - 7 | 1838 | 61 | |
BT015267.1 | - | 6 - 7 | 1838 | 61 | ||
BT132967.1 | - | 6 - 7 | 1977 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |