Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103504.2 | NP_001096974.1 | 9 - 10 | 4742 | 143 | |
NM_001169274.2 | NP_001162745.1 | 8 - 9 | 4115 | 143 | ||
NM_001169275.2 | NP_001162746.1 | 8 - 9 | 4334 | 143 | ||
NM_001272597.2 | NP_001259526.2 | 9 - 10 | 4872 | 143 | ||
NM_001298290.1 | NP_001285219.1 | 9 - 10 | 4140 | 143 | ||
NM_078592.3 | NP_511147.2 | 9 - 10 | 4961 | 143 | ||
GenBank mRNA | AF195496.1 | - | 8 - 9 | 3968 | 143 | |
BT015267.1 | - | 8 - 9 | 3968 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |