Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01813727_m1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813727_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042815.2
NP_001036280.1
19 - 20
3171
62
NM_001201671.1
NP_001188600.1
19 - 20
3171
62
NM_001201673.1
NP_001188602.1
18 - 19
3108
62
NM_001201674.1
NP_001188603.1
18 - 19
3138
62
NM_001201675.1
NP_001188604.1
18 - 19
3084
62
NM_001201679.1
NP_001188608.1
19 - 20
3147
62
NM_001201680.1
NP_001188609.1
18 - 19
3108
62
NM_001201681.1
NP_001188610.1
18 - 19
3084
62
NM_001201682.1
NP_001188611.1
19 - 20
3171
62
NM_001201683.1
NP_001188612.1
19 - 20
3075
62
NM_001201686.1
NP_001188615.1
19 - 20
3147
62
NM_001201687.1
NP_001188616.1
19 - 20
3171
62
NM_001201688.1
NP_001188617.1
18 - 19
3108
62
NM_001201689.1
NP_001188618.1
18 - 19
3084
62
NM_001201693.1
NP_001188622.1
19 - 20
3099
62
NM_001201695.2
NP_001188624.1
19 - 20
3099
62
NM_001201696.1
NP_001188625.1
17 - 18
3075
62
NM_001201697.1
NP_001188626.1
19 - 20
3171
62
NM_001201698.1
NP_001188627.1
18 - 19
3138
62
NM_001201699.1
NP_001188628.1
17 - 18
3075
62
NM_001201700.1
NP_001188629.1
18 - 19
3138
62
NM_001201701.1
NP_001188630.1
17 - 18
3075
62
NM_001201702.1
NP_001188631.1
18 - 19
3138
62
NM_001201703.3
NP_001188632.1
19 - 20
3661
62
NM_001201704.1
NP_001188633.1
17 - 18
3075
62
NM_001201705.1
NP_001188634.1
18 - 19
3066
62
NM_001201706.1
NP_001188635.1
19 - 20
3171
62
NM_001201707.1
NP_001188636.1
19 - 20
3147
62
NM_001201708.1
NP_001188637.1
18 - 19
3108
62
NM_001201709.1
NP_001188638.1
18 - 19
3084
62
NM_001201710.1
NP_001188639.1
15 - 16
2854
62
NM_001201711.1
NP_001188640.1
17 - 18
3009
62
NM_001201712.1
NP_001188641.1
16 - 17
2946
62
NM_001201713.1
NP_001188642.1
16 - 17
2946
62
NM_001201714.1
NP_001188643.1
16 - 17
2922
62
NM_001201715.1
NP_001188644.1
17 - 18
3003
62
NM_001201716.1
NP_001188645.1
18 - 19
3114
62
NM_001201717.1
NP_001188646.1
18 - 19
3138
62
NM_001201718.1
NP_001188647.1
17 - 18
3075
62
NM_001201720.1
NP_001188649.1
18 - 19
3138
62
NM_001201722.1
NP_001188651.1
18 - 19
3138
62
NM_001272690.1
NP_001259619.1
19 - 20
3171
62
NM_001298403.1
NP_001285332.1
19 - 20
3324
62
NM_001298404.1
NP_001285333.1
18 - 19
3135
62
NM_078647.4
NP_523371.2
19 - 20
3171
62
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0