Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258793.3 | NP_001245722.1 | 2 - 3 | 1527 | 72 | |
NM_078656.4 | NP_523380.2 | 2 - 3 | 1527 | 72 | ||
GenBank mRNA | BT021343.1 | - | 1460 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |