Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc16F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078670.3 | NP_523394.1 | 20 - 21 | 8389 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |