Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKLR-17D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103553.3 | NP_001097023.1 | 2 - 3 | 1016 | 71 | |
GenBank mRNA | AY231149.1 | - | 2 - 3 | 1220 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Drosophila melanogaster |