Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dome | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078688.3 | NP_523412.1 | 1 - 2 | 1685 | 75 | |
GenBank mRNA | AJ420377.1 | - | 1 - 2 | 1615 | 75 | |
AY075427.1 | - | 1 - 2 | 1604 | 75 | ||
AY147847.1 | - | 1 - 2 | 1615 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |