Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273041.1 | NP_001259970.1 | 3 - 4 | 314 | 60 | |
NM_164506.4 | NP_722853.1 | 3 - 4 | 314 | 60 | ||
NM_164507.4 | NP_722854.1 | 3 - 4 | 314 | 60 | ||
NM_164508.3 | NP_722855.1 | 3 - 4 | 338 | 60 | ||
GenBank mRNA | BT015189.1 | - | 3 - 4 | 313 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |