Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298769.1 | NP_001285698.1 | 5 - 6 | 1915 | 90 | |
NM_078773.3 | NP_523497.2 | 5 - 6 | 1915 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |