Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078783.3 | NP_523507.2 | 13 - 14 | 4069 | 103 | |
GenBank mRNA | AY069050.1 | - | 13 - 14 | 4035 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |