Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299173.1 | NP_001286102.1 | 2 - 3 | 702 | 94 | |
NM_136189.4 | NP_610033.1 | 2 - 3 | 669 | 94 | ||
GenBank mRNA | AY060391.1 | - | 2 - 3 | 645 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |