Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: step | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042910.3 | NP_001036375.2 | 8 - 9 | 2135 | 63 | |
NM_001169570.2 | NP_001163041.1 | 7 - 8 | 2396 | 63 | ||
NM_001273749.1 | NP_001260678.1 | 9 - 10 | 2864 | 63 | ||
NM_136276.4 | NP_610120.1 | 7 - 8 | 1087 | 63 | ||
GenBank mRNA | AY071322.1 | - | 7 - 8 | 1021 | 63 | |
BT072836.1 | - | 7 - 8 | 1039 | 63 | ||
BT124891.1 | - | 9 - 10 | 2763 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |