Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: az2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136445.3 | NP_610289.2 | 2 - 3 | 843 | 70 | |
GenBank mRNA | BT021301.1 | - | 2 - 3 | 818 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |