Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136471.4 | NP_610315.1 | 1 | 870 | 115 | |
GenBank mRNA | AY071516.1 | - | 1 | 860 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |