Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166187.2 | NP_725600.1 | 1 - 2 | 417 | 74 | |
GenBank mRNA | BT125004.1 | - | 1 - 2 | 247 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |