Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ena | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014536.2 | NP_001014536.1 | 5 - 6 | 1040 | 101 | |
NM_001144237.2 | NP_001137709.1 | 3 - 4 | 1701 | 101 | ||
NM_166329.2 | NP_725857.1 | 5 - 6 | 964 | 101 | ||
NM_166330.2 | NP_725858.1 | 4 - 5 | 1400 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |