Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079990.3 | NP_524729.2 | 6 - 7 | 3905 | 68 | |
GenBank mRNA | AF239997.1 | - | 6 - 7 | 3882 | 68 | |
AJ310804.1 | - | 6 - 7 | 3910 | 68 | ||
BT058000.1 | - | 6 - 7 | 3913 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |