Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259525.1 | NP_001246454.1 | 3 - 4 | 3865 | 118 | |
NM_130462.4 | NP_569729.1 | 3 - 4 | 3816 | 118 | ||
NM_166505.2 | NP_726155.1 | 3 - 4 | 3882 | 118 | ||
GenBank mRNA | BT016135.1 | - | 3 - 4 | 3953 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |