Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103936.2 | NP_001097406.1 | 3 - 4 | 1329 | 111 | |
NM_001169775.2 | NP_001163246.1 | 3 - 4 | 769 | 111 | ||
NM_001169776.2 | NP_001163247.1 | 3 - 4 | 851 | 111 | ||
NM_001169777.2 | NP_001163248.1 | 3 - 4 | 1329 | 111 | ||
NM_001169778.2 | NP_001163249.1 | 3 - 4 | 851 | 111 | ||
NM_001299775.1 | NP_001286704.1 | 3 - 4 | 769 | 111 | ||
NM_137796.3 | NP_611640.3 | 3 - 4 | 769 | 111 | ||
NM_166507.2 | NP_726159.1 | 3 - 4 | 1042 | 111 | ||
NM_166508.2 | NP_726160.1 | 3 - 4 | 1329 | 111 | ||
NM_166509.2 | NP_726161.1 | 4 - 5 | 1192 | 111 | ||
NM_166510.2 | NP_726162.1 | 3 - 4 | 851 | 111 | ||
NM_166511.2 | NP_726163.1 | 3 - 4 | 837 | 111 | ||
GenBank mRNA | AY094731.1 | - | 3 - 4 | 1343 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |