Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259541.1 | NP_001246470.1 | 8 - 9 | 2066 | 74 | |
NM_166566.1 | NP_726260.1 | 8 - 9 | 2030 | 74 | ||
GenBank mRNA | BT004473.1 | - | 8 - 9 | 2031 | 74 | |
BT120028.1 | - | 8 - 9 | 1993 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |