Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nahoda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259545.2 | NP_001246474.1 | 8 - 9 | 3869 | 74 | |
NM_079091.4 | NP_523815.2 | 8 - 9 | 3869 | 74 | ||
NM_166569.3 | NP_726264.1 | 8 - 9 | 3401 | 74 | ||
GenBank mRNA | AY060632.1 | - | 8 - 9 | 3826 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |