Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ReepA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103955.3 | NP_001097425.1 | 4 - 5 | 890 | 66 | |
NM_166574.3 | NP_726269.1 | 5 - 6 | 826 | 66 | ||
NM_166576.3 | NP_726271.1 | 3 - 4 | 930 | 66 | ||
GenBank mRNA | AY094833.1 | - | 3 - 4 | 1707 | 66 | |
BT001599.1 | - | 4 - 5 | 907 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |