Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273317.1 | NP_001260246.1 | 3 - 4 | 1193 | 64 | |
NM_078786.3 | NP_523510.1 | 3 - 4 | 1193 | 64 | ||
GenBank mRNA | AY071690.1 | - | 3 - 4 | 1188 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |