Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078796.3 | NP_523520.2 | 3 - 4 | 716 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |