Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201817.1 | NP_001188746.1 | 10 - 11 | 5493 | 76 | |
NM_001201818.1 | NP_001188747.1 | 10 - 11 | 5496 | 76 | ||
NM_001201819.2 | NP_001188748.1 | 10 - 11 | 5493 | 76 | ||
NM_001259020.2 | NP_001245949.1 | 8 - 9 | 5688 | 76 | ||
NM_001273351.1 | NP_001260280.1 | 9 - 10 | 5625 | 76 | ||
NM_078797.4 | NP_523521.3 | 9 - 10 | 5625 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |