Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dref | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273382.2 | NP_001260311.1 | 3 - 4 | 2145 | 96 | |
NM_078805.4 | NP_523529.1 | 3 - 4 | 2145 | 96 | ||
GenBank mRNA | AB010823.1 | - | 3 - 4 | 1807 | 96 | |
BT099709.1 | - | 3 - 4 | 2130 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |