Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273426.1 | NP_001260355.1 | 1 - 2 | 662 | 79 | |
NM_078815.3 | NP_523539.2 | 1 - 2 | 662 | 79 | ||
GenBank mRNA | AY058306.1 | - | 1 - 2 | 638 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |