Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382179.1 | NP_001369131.1 | 17 - 18 | 9254 | 88 | |
NM_078867.4 | NP_523591.2 | 17 - 18 | 9254 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |