Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078908.3 | NP_523632.2 | 2 - 3 | 601 | 62 | |
GenBank mRNA | AY070898.1 | - | 2 - 3 | 599 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |