Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Eo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078917.3 | NP_523641.2 | 4 - 5 | 752 | 87 | |
GenBank mRNA | AY070986.1 | - | 4 - 5 | 748 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |