Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273833.1 | NP_001260762.1 | 5 - 6 | 2197 | 77 | |
NM_001299227.1 | NP_001286156.1 | 5 - 6 | 2333 | 77 | ||
NM_078924.5 | NP_523648.3 | 5 - 6 | 2197 | 77 | ||
GenBank mRNA | BT001508.1 | - | 5 - 6 | 2176 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |