Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Damm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316416.1 | NP_001303345.1 | 4 - 5 | 719 | 65 | |
NM_078979.3 | NP_523703.2 | 4 - 5 | 719 | 65 | ||
GenBank mRNA | AF281077.1 | - | 4 - 5 | 690 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |