Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4ad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273876.1 | NP_001260805.1 | 5 - 6 | 807 | 66 | |
NM_136536.3 | NP_610380.1 | 7 - 8 | 1212 | 66 | ||
GenBank mRNA | AY061058.1 | - | 7 - 8 | 1185 | 66 | |
BT133029.1 | - | 5 - 6 | 807 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |