Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299307.1 | NP_001286236.1 | 17 - 18 | 6025 | 61 | |
NM_136623.2 | NP_610467.1 | 17 - 18 | 6028 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |