Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273892.1 | NP_001260821.1 | 2 - 3 | 1740 | 96 | |
NM_001299317.1 | NP_001286246.1 | 2 - 3 | 1406 | 96 | ||
NM_136635.3 | NP_610479.1 | 2 - 3 | 1740 | 96 | ||
GenBank mRNA | AF422803.1 | - | 2 - 3 | 1383 | 96 | |
AY069468.1 | - | 2 - 3 | 1685 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |