Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ClC-b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136954.4 | NP_610798.1 | 6 - 7 | 2147 | 72 | |
GenBank mRNA | AY071581.1 | - | 6 - 7 | 2103 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |