Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299867.1 | NP_001286796.1 | 7 - 8 | 3527 | 97 | |
NM_057484.3 | NP_476832.1 | 7 - 8 | 3527 | 97 | ||
NM_166633.2 | NP_726381.1 | 8 - 9 | 3073 | 97 | ||
NM_166634.2 | NP_726382.1 | 8 - 9 | 3083 | 97 | ||
NM_166635.2 | NP_726383.1 | 8 - 9 | 2947 | 97 | ||
NM_166636.2 | NP_726384.1 | 8 - 9 | 2998 | 97 | ||
NM_166637.2 | NP_726385.1 | 7 - 8 | 3527 | 97 | ||
NM_166638.2 | NP_726386.1 | 8 - 9 | 2957 | 97 | ||
NM_166639.2 | NP_726387.1 | 8 - 9 | 3083 | 97 | ||
GenBank mRNA | BT125984.1 | - | 8 - 9 | 3042 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster Inventoried |