Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SiaT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079129.3 | NP_523853.1 | 4 - 5 | 1843 | 95 | |
NM_166684.2 | NP_726474.1 | 5 - 6 | 1223 | 95 | ||
GenBank mRNA | AF218237.1 | - | 3 - 4 | 1225 | 95 | |
AF397532.1 | - | 4 - 5 | 1186 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |