Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143673.3 | NP_651930.2 | 8 - 9 | 1782 | 107 | |
NM_166779.4 | NP_726594.2 | 8 - 9 | 1740 | 107 | ||
GenBank mRNA | BT010000.1 | - | 8 - 9 | 1780 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |