Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc-76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272243.1 | NP_001259172.1 | 3 - 4 | 1745 | 84 | |
NM_001272244.1 | NP_001259173.1 | 3 - 4 | 966 | 84 | ||
NM_130625.3 | NP_569981.1 | 3 - 4 | 1745 | 84 | ||
NM_166927.2 | NP_726792.1 | 4 - 5 | 1635 | 84 | ||
GenBank mRNA | AY069376.1 | - | 3 - 4 | 1715 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |