Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079016.3 | NP_523740.3 | 4 - 5 | 2692 | 73 | |
GenBank mRNA | BT021362.1 | - | 4 - 5 | 2692 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |