Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079049.4 | NP_523773.1 | 2 - 3 | 655 | 64 | |
GenBank mRNA | AY061421.1 | - | 2 - 3 | 595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |