Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299611.1 | NP_001286540.1 | 5 - 6 | 1438 | 61 | |
NM_079054.5 | NP_523778.2 | 5 - 6 | 1438 | 61 | ||
GenBank mRNA | BT128787.1 | - | 5 - 6 | 1422 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |